Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287819.1 | NP_001274748.1 | 26, 27 | 3340 | ||
NM_001287820.1 | NP_001274749.1 | 26, 27 | 3211 | |||
NM_198503.3 | NP_940905.2 | 27, 28 | 3412 | |||
XM_006711294.2 | XP_006711357.1 | 28, 29 | 3907 | |||
XM_006711295.2 | XP_006711358.1 | 27, 28 | 3835 | |||
XM_011509482.2 | XP_011507784.1 | 27, 28 | 3832 | |||
XM_017001181.1 | XP_016856670.1 | 18, 19 | 2422 | |||
XM_017001182.1 | XP_016856671.1 | 27, 28 | 3798 | |||
XM_017001183.1 | XP_016856672.1 | 15, 16 | 1958 | |||
XM_017001184.1 | XP_016856673.1 | 26, 27 | 3777 | |||
GenBank mRNA | AK127807.1 | - | 19, 20 | 2613 | ||
AY359444.1 | - | 27, 28 | 3350 | |||
BC103947.1 | - | 23, 24 | 2663 | |||
BC103948.1 | - | 26, 27 | 3268 | 40009146 | ||
BC103949.1 | - | 26, 27 | 3139 | 40009147 | ||
BC103950.1 | - | 26, 27 | 3268 | 40009148 | ||
BX647852.1 | - | 26, 27 | 3322 | |||
EU251076.1 | - | 27, 28 | 3290 |