Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169.2 | NP_001160.2 | 5 | 759 | ||
XM_011545822.2 | XP_011544124.1 | 5 | 802 | |||
XM_011545823.2 | XP_011544125.1 | 3 | 467 | |||
GenBank mRNA | AB013456.1 | - | 5 | 761 | ||
AF067797.1 | - | 5 | 762 | |||
AK222964.1 | - | 5 | 724 | |||
AK222975.1 | - | 5 | 756 | |||
BC040630.1 | - | 5 | 727 | 5759105 |