Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCKAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207363.2 | NP_997246.2 | 13 | 1420 | ||
NM_207481.3 | NP_997364.3 | 13 | 1420 | |||
XM_005263659.3 | XP_005263716.1 | 12 | 2002 | |||
XM_005263660.3 | XP_005263717.1 | 12 | 1517 | |||
XM_011511097.2 | XP_011509399.1 | 14 | 2190 | |||
XM_011511098.2 | XP_011509400.1 | 14 | 2187 | |||
XM_011511099.2 | XP_011509401.1 | 13 | 1585 | |||
XM_011511100.2 | XP_011509402.1 | 13 | 1683 | |||
XM_011511101.1 | XP_011509403.1 | 11 | 1093 | |||
XM_011511102.2 | XP_011509404.1 | 12 | 5519 | |||
XM_011511103.2 | XP_011509405.1 | 10 | 1321 | |||
XM_011511104.1 | XP_011509406.1 | 10 | 972 | |||
XM_011511105.2 | XP_011509407.1 | 10 | 1763 | |||
XM_017003974.1 | XP_016859463.1 | 13 | 1313 | |||
XM_017003975.1 | XP_016859464.1 | 12 | 1615 | |||
XM_017003976.1 | XP_016859465.1 | 14 | 2211 | |||
XM_017003977.1 | XP_016859466.1 | 11 | 2121 | |||
XM_017003978.1 | XP_016859467.1 | 7 | 1531 | |||
XM_017003979.1 | XP_016859468.1 | 13 | 2060 | |||
XM_017003980.1 | XP_016859469.1 | 10 | 1321 | |||
GenBank mRNA | AY946007.1 | - | 8 | 842 | ||
AY946008.1 | - | 8 | 842 |