Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRSS48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183375.2 | NP_899231.2 | 5 | 730 | ||
XM_011531921.2 | XP_011530223.1 | 4 | 1176 | |||
XM_011531922.2 | XP_011530224.1 | 3 | 992 | |||
GenBank mRNA | BC121109.1 | - | 5 | 734 | 40120369 | |
BC121110.1 | - | 3 | 305 | 40120371 |