Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSANTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042690.1 | NP_001036155.1 | 1 | 467 | ||
XM_011513465.2 | XP_011511767.1 | 4 | 1729 | |||
XM_011513466.2 | XP_011511768.1 | 2 | 3994 | |||
XM_011513467.2 | XP_011511769.1 | 1 | 5779 | |||
XM_017008169.1 | XP_016863658.1 | 2 | 3172 | |||
GenBank mRNA | AK126094.1 | - | 1 | 467 | ||
BC043530.1 | - | 4 | 1088 |