Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC10A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197965.2 | NP_932069.1 | 4, 5 | 906 | ||
GenBank mRNA | AJ583502.1 | - | 4, 5 | 758 | ||
BC107051.1 | - | 4, 5 | 758 | 40004572 | ||
BC107052.1 | - | 4, 5 | 758 | 40004575 | ||
EF437223.1 | - | 4, 5 | 906 |