Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000040.1 | NP_000031.1 | 2 | 67 | ||
GenBank mRNA | BC027977.1 | - | 2 | 65 | 5229117 | |
BC121081.1 | - | 2 | 67 | 40119733 | ||
BC134419.1 | - | 2 | 67 | 40119734 | ||
M28613.1 | - | 1 | 51 | |||
M28614.1 | - | 1 | 51 | |||
X00567.1 | - | 1 | 21 | |||
X01388.1 | - | 2 | 92 |