Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6orf52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145020.1 | NP_001138492.1 | 3 | 379 | ||
NR_026736.1 | 2 | 279 | ||||
NR_026737.1 | 2 | 297 | ||||
XM_006715088.3 | XP_006715151.1 | 3 | 548 | |||
XM_011514572.2 | XP_011512874.1 | 2 | 344 | |||
XM_011514574.2 | XP_011512876.1 | 2 | 344 | |||
XM_011514575.2 | XP_011512877.1 | 3 | 530 | |||
GenBank mRNA | AI621292.1 | - | 5 | 411 | ||
AI809192.1 | - | 3 | 237 | |||
AW183566.1 | - | 3 | 239 | |||
AW293245.1 | - | 3 | 253 | |||
BC016820.1 | - | 2 | 193 | |||
BX280692.1 | - | 2 | 279 |