Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM255B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182614.2 | NP_872420.1 | 9 | 957 | ||
XM_005272413.3 | XP_005272470.1 | 10 | 1076 | |||
XM_005272415.2 | XP_005272472.1 | 10 | 1058 | |||
XM_006719986.2 | XP_006720049.1 | 7 | 752 | |||
XM_017020558.1 | XP_016876047.1 | 11 | 1148 | |||
XM_017020559.1 | XP_016876048.1 | 12 | 2357 | |||
XM_017020560.1 | XP_016876049.1 | 8 | 920 | |||
GenBank mRNA | BC018995.2 | - | 9 | 957 | 4300679 |