Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM255B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182614.2 | NP_872420.1 | 4 | 343 | ||
XM_005272413.3 | XP_005272470.1 | 4 | 369 | |||
XM_005272415.2 | XP_005272472.1 | 4 | 343 | |||
XM_017020558.1 | XP_016876047.1 | 4 | 369 | |||
XM_017020559.1 | XP_016876048.1 | 5 | 1578 | |||
XM_017020560.1 | XP_016876049.1 | 3 | 306 | |||
GenBank mRNA | BC018995.2 | - | 4 | 343 | 4300679 |