Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGFBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000597.2 | NP_000588.2 | 2 | 747 | ||
NM_001313990.1 | NP_001300919.1 | 2 | 305 | |||
NM_001313992.1 | NP_001300921.1 | 2 | 624 | |||
NM_001313993.1 | NP_001300922.1 | 2 | 363 | |||
GenBank mRNA | A09809.1 | - | 2 | 746 | ||
AB209509.1 | - | 2 | 722 | |||
AL559513.3 | - | 2 | 754 | |||
BC004312.1 | - | 2 | 721 | 3627826 | ||
BC009902.1 | - | 2 | 723 | 3509749 | ||
BC012769.1 | - | 2 | 721 | 3689990 | ||
BC071967.1 | - | 2 | 734 | 6650239 | ||
BE735562.1 | - | 2 | 305 | |||
CN431340.1 | - | 2 | 363 | |||
CV810939.1 | - | 2 | 681 | |||
M35410.1 | - | 2 | 690 | |||
X16302.1 | - | 2 | 744 |