Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIPAL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099287.1 | NP_001092757.1 | 6 | 1443 | ||
NM_001172292.1 | NP_001165763.1 | 5 | 1386 | |||
XM_011534552.1 | XP_011532854.1 | 6 | 1355 | |||
GenBank mRNA | AK026158.1 | - | 6 | 1247 | ||
AK296972.1 | - | 5 | 1386 | |||
BC105708.1 | - | 5 | 980 | |||
BC105709.1 | - | 5 | 975 | |||
BC105710.1 | - | 5 | 980 |