Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000041.3 | NP_000032.1 | 3 | 293 | ||
NM_001302688.1 | NP_001289617.1 | 3 | 375 | |||
NM_001302689.1 | NP_001289618.1 | 3 | 224 | |||
NM_001302690.1 | NP_001289619.1 | 3 | 324 | |||
NM_001302691.1 | NP_001289620.1 | 3 | 308 | |||
GenBank mRNA | AK314898.1 | - | 3 | 246 | ||
BC003557.1 | - | 3 | 222 | 3355712 | ||
BI550631.1 | - | 3 | 270 | |||
BM042032.1 | - | 3 | 308 | |||
BU848796.1 | - | 260 | ||||
CA771535.1 | - | 2 | 442 | |||
DB486513.1 | - | 3 | 225 | |||
HY056112.1 | - | 3 | 293 | |||
HY074384.1 | - | 3 | 224 | |||
K00396.1 | - | 3 | 237 | |||
KU177910.1 | - | 2 | 177 | |||
KU177911.1 | - | 2 | 177 | |||
KU177912.1 | - | 2 | 177 | |||
KU177913.1 | - | 2 | 177 | |||
M12529.1 | - | 3 | 238 | |||
Z70760.1 | - | 2 | 214 |