Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000041.3 | NP_000032.1 | 3, 4 | 362 | ||
NM_001302688.1 | NP_001289617.1 | 3, 4 | 444 | |||
NM_001302689.1 | NP_001289618.1 | 3, 4 | 293 | |||
NM_001302690.1 | NP_001289619.1 | 3, 4 | 393 | |||
NM_001302691.1 | NP_001289620.1 | 3, 4 | 377 | |||
GenBank mRNA | AK314898.1 | - | 3, 4 | 315 | ||
BC003557.1 | - | 3, 4 | 291 | 3355712 | ||
BI550631.1 | - | 3, 4 | 339 | |||
BM042032.1 | - | 3, 4 | 377 | |||
BU848796.1 | - | 329 | ||||
CA771535.1 | - | 2, 3 | 511 | |||
DB486513.1 | - | 3, 4 | 294 | |||
HY056112.1 | - | 3, 4 | 362 | |||
K00396.1 | - | 3, 4 | 306 | |||
KU177910.1 | - | 2, 3 | 246 | |||
KU177911.1 | - | 2, 3 | 246 | |||
M12529.1 | - | 3, 4 | 307 | |||
X00199.1 | - | 1, 2 | 57 |