Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBPJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005349.3 | NP_005340.2 | 6 | 653 | ||
NM_015874.4 | NP_056958.3 | 5 | 600 | |||
NM_203283.2 | NP_976028.1 | 5 | 537 | |||
NM_203284.2 | NP_976029.1 | 6 | 783 | |||
XM_005248161.3 | XP_005248218.1 | 6 | 667 | |||
XM_011513840.2 | XP_011512142.1 | 6 | 1059 | |||
XM_017008170.1 | XP_016863659.1 | 5 | 456 | |||
XM_017008171.1 | XP_016863660.1 | 5 | 435 | |||
XM_017008172.1 | XP_016863661.1 | 6 | 598 | |||
XM_017008173.1 | XP_016863662.1 | 6 | 876 | |||
XM_017008174.1 | XP_016863663.1 | 6 | 692 | |||
XM_017008175.1 | XP_016863664.1 | 6 | 608 | |||
GenBank mRNA | AK302230.1 | - | 5 | 390 | ||
AK303244.1 | - | 5 | 449 | |||
AK311419.1 | - | 5 | 436 | |||
BC020780.1 | - | 5 | 537 | 4718839 | ||
BC064976.1 | - | 6 | 558 | 6152924 | ||
BQ216033.1 | - | 5 | 581 |