Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAET1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001788.3 | NP_001001788.2 | 5 | 1115 | ||
NR_130110.1 | 5 | 1015 | ||||
GenBank mRNA | AY172579.1 | - | 5 | 1041 | ||
AY172580.1 | - | 5 | 873 |