Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL1R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000877.3 | NP_000868.1 | 8 | 1065 | ||
NM_001288706.1 | NP_001275635.1 | 8 | 978 | |||
NM_001320978.1 | NP_001307907.1 | 8 | 978 | |||
NM_001320980.1 | NP_001307909.1 | 8 | 1119 | |||
NM_001320981.1 | NP_001307910.1 | 8 | 960 | |||
NM_001320982.1 | NP_001307911.1 | 8 | 902 | |||
NM_001320983.1 | NP_001307912.1 | 6 | 753 | |||
NM_001320984.1 | NP_001307913.1 | 6 | 630 | |||
NM_001320985.1 | NP_001307914.1 | 6 | 651 | |||
XM_005263930.4 | XP_005263987.1 | 8 | 927 | |||
XM_005263934.4 | XP_005263991.1 | 9 | 1032 | |||
XM_011511115.2 | XP_011509417.1 | 9 | 1024 | |||
XM_011511116.1 | XP_011509418.1 | 8 | 1002 | |||
XM_011511117.1 | XP_011509419.1 | 8 | 997 | |||
XM_011511118.2 | XP_011509420.1 | 8 | 928 | |||
XM_017003989.1 | XP_016859478.1 | 8 | 1125 | |||
GenBank mRNA | AK314433.1 | - | 8 | 978 | ||
BC067506.1 | - | 7 | 799 | 7002107 | ||
BC067507.1 | - | 7 | 801 | 7002109 | ||
BC067508.1 | - | 7 | 801 | 7002111 | ||
BC075062.2 | - | 7 | 781 | 30915370 | ||
BC075063.2 | - | 7 | 781 | 30915586 | ||
CD013912.1 | - | 6 | 622 | |||
CD013913.1 | - | 6 | 643 | |||
M27492.1 | - | 7 | 819 | |||
X16896.1 | - | 8 | 1045 |