Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168298.1 | NP_001161770.1 | 4 | 834 | ||
NM_001557.3 | NP_001548.1 | 3 | 1042 | |||
XM_005246530.3 | XP_005246587.1 | 4 | 814 | |||
XM_017003990.1 | XP_016859479.1 | 4 | 794 | |||
XM_017003991.1 | XP_016859480.1 | 2 | 705 | |||
XM_017003992.1 | XP_016859481.1 | 2 | 975 | |||
GenBank mRNA | AK290906.1 | - | 3 | 940 | ||
AK312664.1 | - | 3 | 933 | |||
BC037961.1 | - | 3 | 996 | 5752441 | ||
L19593.1 | - | 3 | 1032 | |||
M73969.1 | - | 1 | 649 | |||
M94582.1 | - | 2 | 676 |