Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000572.2 | NP_000563.1 | 2 | 244 | ||
GenBank mRNA | AF043333.1 | - | 2 | 131 | ||
AY029171.1 | - | 2 | 185 | |||
BC104252.1 | - | 2 | 241 | 40035920 | ||
BC104253.1 | - | 2 | 241 | 40035922 | ||
CR541993.1 | - | 2 | 185 | |||
CR542028.1 | - | 2 | 185 | |||
M57627.1 | - | 2 | 215 |