Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198098.2 | NP_932766.1 | 1 | 448 | ||
GenBank mRNA | AB451275.1 | - | 1 | 338 | ||
AB451402.1 | - | 1 | 338 | |||
AK298091.1 | - | 2 | 350 | |||
AK298313.1 | - | 2 | 209 | |||
AK300995.1 | - | 2 | 260 | |||
AK304684.1 | - | 2 | 302 | |||
AK312656.1 | - | 1 | 395 | |||
AU122128.1 | - | 1 | 395 | |||
BC022486.1 | - | 1 | 395 | 4799283 | ||
BI837828.1 | - | 1 | 385 | |||
M77829.1 | - | 1 | 376 | |||
S73482.1 | - | 1 | 376 | |||
U41517.1 | - | 1 | 371 |