Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL11RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142784.2 | NP_001136256.1 | 5 | 455 | ||
NR_052010.1 | 5 | 509 | ||||
GenBank mRNA | AK075530.1 | - | 5 | 487 | ||
AK094689.1 | - | 5 | 1762 | |||
AK292073.1 | - | 5 | 489 | |||
BC003110.1 | - | 5 | 471 | 3502059 | ||
BT009864.1 | - | 4 | 406 | |||
DA917660.1 | - | 5 | 455 | |||
DC424068.1 | - | 5 | 509 | |||
U32324.1 | - | 5 | 467 | |||
Z38102.1 | - | 4 | 411 | |||
Z46595.1 | - | 4 | 400 |