Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002188.2 | NP_002179.2 | 3 | 308 | ||
GenBank mRNA | BC096138.3 | - | 2 | 1365 | 40002676 | |
BC096139.1 | - | 3 | 308 | 40002680 | ||
BC096140.3 | - | 2 | 1363 | 40002684 | ||
BC096141.1 | - | 3 | 308 | 40002686 | ||
L06801.1 | - | 3 | 296 | |||
X69079.1 | - | 3 | 308 |