Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ILF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267809.1 | NP_001254738.1 | 7 | 570 | ||
NM_004515.3 | NP_004506.2 | 7 | 551 | |||
GenBank mRNA | AK223326.1 | - | 7 | 471 | ||
AK302214.1 | - | 7 | 514 | |||
AK310615.1 | - | 7 | 534 | |||
AK313364.1 | - | 7 | 510 | |||
BC000382.2 | - | 7 | 476 | 2820505 | ||
BM800544.1 | - | 7 | 526 | |||
DC412988.1 | - | 7 | 551 | |||
HQ322261.1 | - | 6 | 445 | |||
HQ322262.1 | - | 6 | 445 | |||
U10323.1 | - | 7 | 484 |