Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMPDH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000883.3 | NP_000874.2 | 6, 7 | 591 | ||
NM_001102605.1 | NP_001096075.1 | 5, 6 | 561 | |||
NM_001142573.1 | NP_001136045.1 | 3, 4 | 361 | |||
NM_001142574.1 | NP_001136046.1 | 3, 4 | 361 | |||
NM_001142576.1 | NP_001136048.1 | 5, 6 | 492 | |||
NM_001304521.1 | NP_001291450.1 | 3, 4 | 389 | |||
NM_183243.2 | NP_899066.1 | 4, 5 | 483 | |||
XM_017012168.1 | XP_016867657.1 | 5, 6 | 561 | |||
XM_017012169.1 | XP_016867658.1 | 4, 5 | 483 | |||
XM_017012170.1 | XP_016867659.1 | 3, 4 | 390 | |||
XM_017012171.1 | XP_016867660.1 | 5, 6 | 679 | |||
XM_017012172.1 | XP_016867661.1 | 5, 6 | 679 | |||
XM_017012173.1 | XP_016867662.1 | 5, 6 | 561 | |||
GenBank mRNA | AK054667.1 | - | 4, 5 | 483 | ||
AK092452.1 | - | 5, 6 | 528 | |||
AK122994.1 | - | 5, 6 | 560 | |||
AK131294.1 | - | 3, 4 | 308 | |||
AK293413.1 | - | 3, 4 | 333 | |||
AK297943.1 | - | 3, 4 | 333 | |||
BC033622.1 | - | 4, 5 | 474 | 5210847 | ||
BX406207.2 | - | 4, 5 | 486 | |||
CD014008.1 | - | 4, 5 | 275 | |||
CD014010.1 | - | 5, 6 | 375 | |||
CR933672.1 | - | 527 | ||||
DA980797.1 | - | 4, 5 | 483 | |||
DC299597.1 | - | 3, 4 | 361 |