Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317384.1 | NP_001304313.1 | 5 | 1025 | ||
NM_001317387.1 | NP_001304316.1 | 4 | 944 | |||
NM_001650.5 | NP_001641.1 | 5 | 1025 | |||
NM_004028.4 | NP_004019.1 | 4 | 1152 | |||
XM_011525942.2 | XP_011524244.1 | 5 | 1067 | |||
GenBank mRNA | AB128929.1 | - | 6 | 555 | ||
AB209156.1 | - | 3 | 558 | |||
AK222684.1 | - | 5 | 937 | |||
AK295069.1 | - | 6 | 836 | |||
BC022286.1 | - | 5 | 890 | 4717755 | ||
D63412.1 | - | 5 | 890 | |||
HQ901095.1 | - | 5 | 999 | |||
KF055862.1 | - | 4 | 789 | |||
U63622.1 | - | 5 | 909 | |||
U63623.1 | - | 4 | 813 |