Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INSL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001265587.1 | NP_001252516.1 | 3 | 520 | ||
NM_005543.3 | NP_005534.2 | 2 | 425 | |||
GenBank mRNA | AK302780.1 | - | 2 | 367 | ||
AY082014.1 | - | 3 | 520 | |||
BC032810.1 | - | 2 | 355 | |||
BC040435.1 | - | 2 | 355 | |||
BC053345.1 | - | 2 | 355 | |||
BC061886.1 | - | 2 | 378 | |||
BC071706.1 | - | 2 | 358 | 5167223 | ||
BC106721.1 | - | 2 | 357 | 40014433 | ||
BC106722.1 | - | 2 | 362 | 40014435 | ||
BC107045.2 | - | 1 | 51 | |||
BX334591.2 | - | 2 | 368 | |||
S72482.1 | - | 2 | 364 |