Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002202.2 | NP_002193.2 | 2, 3 | 764 | ||
GenBank mRNA | AK290742.1 | - | 2, 3 | 482 | ||
BC031213.1 | - | 2, 3 | 765 | 5269369 | ||
S70721.1 | - | 2, 3 | 221 | |||
U07559.1 | - | 2, 3 | 455 |