Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000419.4 | NP_000410.2 | 16 | 1709 | ||
XM_011524749.1 | XP_011523051.1 | 16 | 1613 | |||
XM_011524750.1 | XP_011523052.1 | 16 | 1613 | |||
GenBank mRNA | AB209552.1 | - | 4 | 376 | ||
AK303081.1 | - | 10 | 1096 | |||
AK315335.1 | - | 16 | 1613 | |||
BC117443.1 | - | 16 | 1607 | 40125994 | ||
BC126442.1 | - | 16 | 1607 | 8992158 | ||
J02764.1 | - | 16 | 1582 | |||
M34480.1 | - | 16 | 1613 | |||
X06831.1 | - | 5 | 408 |