Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000044.3 | NP_000035.2 | 2 | 2818 | ||
NM_001011645.2 | NP_001011645.1 | 2 | 269 | |||
GenBank mRNA | AF162704.1 | - | 4 | 1664 | ||
BC132975.1 | - | 2 | 2148 | 40146997 | ||
FJ235916.1 | - | 2 | 2033 | |||
FJ235917.1 | - | 2 | 2030 | |||
FJ235918.1 | - | 2 | 2030 | |||
FJ235919.1 | - | 2 | 2033 | |||
GU208210.1 | - | 2 | 1721 | |||
HQ692823.1 | - | 2 | 107 | |||
KJ534771.1 | - | 2 | 1700 | |||
L29496.1 | - | 2 | 1145 | |||
M20132.1 | - | 2 | 2062 | |||
M21748.1 | - | 2 | 1856 | |||
M23263.1 | - | 2 | 2228 | |||
M34233.1 | - | 4 | 1769 | |||
M73069.1 | - | 3 | 1727 |