Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002213.4 | NP_002204.2 | 14, 15 | 2604 | ||
XM_005247436.2 | XP_005247493.1 | 13, 14 | 2747 | |||
XM_006713630.2 | XP_006713693.1 | 13, 14 | 2702 | |||
XM_017006351.1 | XP_016861840.1 | 14, 15 | 2375 | |||
XM_017006352.1 | XP_016861841.1 | 14, 15 | 2314 | |||
XM_017006353.1 | XP_016861842.1 | 14, 15 | 2346 | |||
XM_017006354.1 | XP_016861843.1 | 11, 12 | 2489 | |||
XM_017006355.1 | XP_016861844.1 | 13, 14 | 2224 | |||
GenBank mRNA | AK054968.1 | - | 3, 4 | 1452 | ||
AK130168.1 | - | 6, 7 | 1003 | |||
AK309551.1 | - | 6, 7 | 1090 | |||
AK315429.1 | - | 14, 15 | 2340 | |||
BC006541.2 | - | 14, 15 | 2596 | 2958666 | ||
J05633.1 | - | 14, 15 | 2345 | |||
M35011.1 | - | 14, 15 | 2328 | |||
X53002.1 | - | 14, 15 | 2635 |