Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITIH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002216.2 | NP_002207.2 | 14 | 1909 | ||
GenBank mRNA | AK225277.1 | - | 16 | 1772 | ||
AK225329.1 | - | 14 | 1814 | |||
AK291379.1 | - | 14 | 1857 | |||
AK297334.1 | - | 9 | 1100 | |||
BC132685.1 | - | 14 | 1796 | 40146707 | ||
M18193.1 | - | 4 | 600 | |||
M33033.1 | - | 4 | 598 | |||
X07173.1 | - | 14 | 1810 |