Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JARID2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267040.1 | NP_001253969.1 | 7 | 1958 | ||
NM_004973.3 | NP_004964.2 | 7 | 2458 | |||
XM_005249089.3 | XP_005249146.1 | 6 | 1796 | |||
XM_011514579.2 | XP_011512881.1 | 9 | 2647 | |||
XM_011514584.2 | XP_011512886.1 | 5 | 1712 | |||
XM_017010833.1 | XP_016866322.1 | 9 | 2602 | |||
XM_017010834.1 | XP_016866323.1 | 6 | 2316 | |||
XM_017010835.1 | XP_016866324.1 | 5 | 1691 | |||
GenBank mRNA | AK292861.1 | - | 7 | 2304 | ||
AK299349.1 | - | 6 | 1872 | |||
AK302624.1 | - | 6 | 1783 | |||
AK303610.1 | - | 7 | 1901 | |||
BC046246.1 | - | 7 | 2175 | 4520786 | ||
U57592.1 | - | 7 | 2137 |