Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC172 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198515.2 | NP_940917.1 | 8 | 939 | ||
XM_006717820.3 | XP_006717883.1 | 8 | 1234 | |||
XM_017016206.1 | XP_016871695.1 | 8 | 857 | |||
GenBank mRNA | AJ457798.1 | - | 7 | 921 | ||
BC044830.1 | - | 8 | 939 | 5166167 |