Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031886.2 | NP_114092.2 | 2 | 1582 | ||
GenBank mRNA | AF315818.1 | - | 2 | 1263 | ||
AJ310479.1 | - | 2 | 1582 | |||
AY779768.1 | - | 2 | 1255 | |||
BC140906.1 | - | 2 | 1376 | 9056718 |