Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286449.1 | NP_001273378.1 | 8 | 884 | ||
NM_198524.2 | NP_940926.1 | 8 | 661 | |||
XM_005254359.4 | XP_005254416.1 | 8 | 831 | |||
XM_005254361.3 | XP_005254418.1 | 7 | 823 | |||
XM_011521530.2 | XP_011519832.1 | 8 | 831 | |||
XM_017022160.1 | XP_016877649.1 | 8 | 831 | |||
XM_017022161.1 | XP_016877650.1 | 6 | 578 | |||
XM_017022162.1 | XP_016877651.1 | 8 | 710 | |||
XM_017022163.1 | XP_016877652.1 | 7 | 923 | |||
XM_017022164.1 | XP_016877653.1 | 8 | 882 | |||
XM_017022166.1 | XP_016877655.1 | 5 | 602 | |||
XM_017022167.1 | XP_016877656.1 | 8 | 882 | |||
GenBank mRNA | AK294960.1 | - | 8 | 870 | ||
AL832050.1 | - | 7 | 740 | |||
BC028119.1 | - | 8 | 639 | 5172473 | ||
KU178902.1 | - | 8 | 634 | |||
KU178903.1 | - | 8 | 857 |