Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198525.2 | NP_940927.2 | 18 | 3686 | ||
XM_011521531.2 | XP_011519833.1 | 18 | 3796 | |||
GenBank mRNA | AY358384.1 | - | 14 | 2255 | ||
BC040878.1 | - | 6 | 985 | |||
BC104044.1 | - | 14 | 2228 | 40026959 | ||
BC112271.1 | - | 13 | 2119 | 8327739 | ||
BC112273.1 | - | 13 | 2119 | 8327741 | ||
BC143461.1 | - | 13 | 2115 | 9051969 |