Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CERKL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030311.2 | NP_001025482.1 | 11 | 1378 | ||
NM_001030312.2 | NP_001025483.1 | 7 | 961 | |||
NM_001030313.2 | NP_001025484.1 | 8 | 1093 | |||
NM_001160277.1 | NP_001153749.1 | 10 | 1246 | |||
NM_201548.4 | NP_963842.1 | 10 | 1300 | |||
NR_027689.1 | 8 | 1104 | ||||
NR_027690.1 | 9 | 1236 | ||||
GenBank mRNA | AJ640141.1 | - | 10 | 1217 | ||
AJ697855.1 | - | 8 | 2005 | |||
AJ697856.1 | - | 9 | 1135 | |||
AJ697858.1 | - | 7 | 993 | |||
AK129976.1 | - | 7 | 543 | |||
AK293844.1 | - | 10 | 1247 | |||
AY357073.1 | - | 10 | 1199 | |||
AY690329.1 | - | 11 | 1277 | |||
AY690330.1 | - | 7 | 860 | |||
AY690331.1 | - | 8 | 992 | |||
AY690332.1 | - | 8 | 1003 | |||
AY690333.1 | - | 9 | 1135 | |||
BC137498.1 | - | 10 | 1226 | 9021498 | ||
BC137499.1 | - | 10 | 1226 | 9021499 | ||
HQ426666.1 | - | 10 | 1106 | |||
HQ426667.1 | - | 9 | 1042 | |||
HQ426668.1 | - | 8 | 910 | |||
HQ426669.1 | - | 8 | 899 | |||
HQ426670.1 | - | 7 | 767 | |||
HQ426671.1 | - | 11 | 1106 | |||
HQ426672.1 | - | 10 | 1028 | |||
HQ426673.1 | - | 9 | 964 | |||
HQ426674.1 | - | 8 | 832 | |||
HQ426675.1 | - | 8 | 821 | |||
HQ426676.1 | - | 7 | 689 | |||
HQ426677.1 | - | 8 | 636 | |||
HQ426678.1 | - | 11 | 1353 | |||
HQ426679.1 | - | 8 | 1658 | |||
JF290420.1 | - | 10 | 1186 |