Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNJ8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004982.3 | NP_004973.1 | 3 | 1385 | ||
XM_005253358.4 | XP_005253415.1 | 4 | 1354 | |||
XM_017019283.1 | XP_016874772.1 | 4 | 1289 | |||
XM_017019284.1 | XP_016874773.1 | 4 | 1360 | |||
GenBank mRNA | AK291821.1 | - | 4 | 1193 | ||
BC000544.2 | - | 3 | 1245 | 3161776 | ||
D50312.1 | - | 3 | 1309 |