Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198581.2 | NP_940983.2 | 4, 5 | 1001 | ||
XM_006712519.2 | XP_006712582.1 | 4, 5 | 1008 | |||
GenBank mRNA | AB111887.1 | - | 3, 4 | 447 | ||
AK127776.1 | - | 1, 2 | 70 | |||
AK131416.1 | - | 4, 5 | 987 | |||
BC155540.1 | - | 3, 4 | 447 | |||
BC167154.1 | - | 3, 4 | 447 |