Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004522.2 | NP_004513.1 | 15 | 2042 | ||
NR_111932.1 | 8 | 1064 | ||||
XM_011511157.2 | XP_011509459.1 | 8 | 1123 | |||
XM_017004062.1 | XP_016859551.1 | 15 | 2033 | |||
GenBank mRNA | AB011103.1 | - | 15 | 2015 | ||
AB073396.1 | - | 3 | 346 | |||
AF010146.1 | - | 5 | 586 | |||
BC110287.1 | - | 8 | 994 | 5165262 | ||
BC127772.1 | - | 12 | 1366 |