Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIR2DL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080770.1 | NP_001074239.1 | 4 | 642 | ||
NM_001080772.1 | NP_001074241.1 | 4 | 642 | |||
NM_002255.5 | NP_002246.5 | 4 | 642 | |||
GenBank mRNA | AF002256.1 | - | 3 | 313 | ||
AF002979.1 | - | 4 | 602 | |||
AF002980.1 | - | 4 | 614 | |||
AF002981.1 | - | 4 | 614 | |||
AF002982.1 | - | 4 | 614 | |||
AF034771.1 | - | 4 | 608 | |||
AF034772.1 | - | 4 | 608 | |||
AF034773.1 | - | 4 | 608 | |||
AF276292.1 | - | 4 | 602 | |||
AF285436.1 | - | 4 | 629 | |||
AK309531.1 | - | 5 | 878 | |||
AY052496.1 | - | 1 | 173 | |||
AY052497.1 | - | 1 | 173 | |||
AY052498.1 | - | 1 | 173 | |||
AY223512.2 | - | 2 | 529 | |||
AY223513.1 | - | 2 | 529 | |||
AY223514.1 | - | 2 | 529 | |||
AY223515.1 | - | 2 | 529 | |||
AY250088.1 | - | 2 | 529 | |||
AY359817.1 | - | 4 | 613 | |||
AY601812.1 | - | 4 | 596 | |||
AY789058.1 | - | 4 | 596 | |||
AY789059.1 | - | 4 | 596 | |||
AY789060.1 | - | 4 | 596 | |||
AY789061.1 | - | 4 | 596 | |||
AY789062.1 | - | 4 | 596 | |||
BC041611.1 | - | 4 | 643 | 5485491 | ||
BM918119.1 | - | 642 | ||||
DQ266438.1 | - | 4 | 605 | |||
DQ272466.1 | - | 2 | 331 | |||
EU194340.1 | - | 4 | 602 | |||
EU194341.1 | - | 3 | 387 | |||
EU194342.1 | - | 4 | 602 | |||
U71199.1 | - | 4 | 614 | |||
U73394.1 | - | 4 | 614 | |||
X97229.1 | - | 4 | 607 | |||
X99479.1 | - | 4 | 602 | |||
X99481.1 | - | 3 | 328 | |||
Y13054.1 | - | 4 | 602 |