Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304448.1 | NP_001291377.1 | 3 | 511 | ||
NM_002259.4 | NP_002250.1 | 2 | 268 | |||
NM_007328.3 | NP_015567.1 | 2 | 268 | |||
NM_213657.2 | NP_998822.1 | 3 | 442 | |||
NM_213658.2 | NP_998823.1 | 3 | 442 | |||
GenBank mRNA | AF461812.1 | - | 2 | 244 | ||
AK310992.1 | - | 3 | 511 | |||
BC012550.1 | - | 3 | 446 | 4042168 | ||
BC053840.1 | - | 3 | 211 | 5484921 | ||
DB464226.1 | - | 2 | 268 | |||
X54867.1 | - | 2 | 218 | |||
X54868.1 | - | 2 | 218 |