Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199352.3 | NP_955384.3 | 4 | 777 | ||
XM_017017684.1 | XP_016873173.1 | 8 | 1946 | |||
XM_017017685.1 | XP_016873174.1 | 9 | 2074 | |||
XM_017017686.1 | XP_016873175.1 | 8 | 2382 | |||
XM_017017687.1 | XP_016873176.1 | 7 | 2397 | |||
XM_017017688.1 | XP_016873177.1 | 4 | 1335 | |||
XM_017017689.1 | XP_016873178.1 | 5 | 1247 | |||
XM_017017690.1 | XP_016873179.1 | 7 | 1150 | |||
XM_017017691.1 | XP_016873180.1 | 5 | 603 | |||
XM_017017692.1 | XP_016873181.1 | 4 | 774 | |||
XM_017017693.1 | XP_016873182.1 | 3 | 365 | |||
XM_017017694.1 | XP_016873183.1 | 3 | 1204 | |||
XM_017017695.1 | XP_016873184.1 | 8 | 2487 | |||
XM_017017696.1 | XP_016873185.1 | 5 | 906 | |||
GenBank mRNA | AK091990.1 | - | 5 | 583 | ||
AK127610.1 | - | 4 | 652 | |||
AY437532.1 | - | 4 | 777 | |||
AY517500.1 | - | 5 | 583 | |||
BC101315.1 | - | 4 | 772 | |||
BC101316.1 | - | 4 | 777 |