Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC184 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013635.3 | NP_001013657.3 | 1 | 657 | ||
GenBank mRNA | AK056003.1 | - | 1 | 624 | ||
AK056783.1 | - | 1 | 131 | |||
BC036801.1 | - | 1 | 608 | 5728402 |