Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002276.4 | NP_002267.2 | 6 | 1166 | ||
GenBank mRNA | AB041267.1 | - | 5 | 594 | ||
AB041268.1 | - | 5 | 593 | |||
AK313261.1 | - | 6 | 1085 | |||
BC002539.2 | - | 6 | 1070 | 3138504 | ||
BC007628.1 | - | 6 | 1070 | 3504206 | ||
BC010409.2 | - | 6 | 1077 | 3689178 | ||
BC067744.1 | - | 6 | 1085 | 30332499 | ||
BC084574.1 | - | 6 | 1068 | 6730698 | ||
Y00503.1 | - | 6 | 1056 |