Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16orf86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012984.2 | NP_001013002.2 | 4 | 1058 | ||
XM_005255952.4 | XP_005256009.1 | 4 | 1093 | |||
GenBank mRNA | AK123796.1 | - | 2 | 1509 | ||
BC064498.1 | - | 4 | 1104 | |||
BC082249.1 | - | 4 | 1104 | |||
CA432406.1 | - | 1 | 226 |