Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMIGD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319942.1 | NP_001306871.1 | 4 | 709 | ||
NM_206832.2 | NP_996663.1 | 4 | 709 | |||
XM_011524787.1 | XP_011523089.1 | 4 | 723 | |||
GenBank mRNA | AK172838.1 | - | 4 | 709 | ||
AK290266.1 | - | 4 | 682 | |||
AY358153.1 | - | 4 | 697 | |||
BC137201.1 | - | 4 | 693 | 9021198 | ||
BC137202.1 | - | 4 | 693 | 9021199 |