Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF790 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242800.1 | NP_001229729.1 | 5 | 496 | ||
NM_001242801.1 | NP_001229730.1 | 5 | 674 | |||
NM_001242802.1 | NP_001229731.1 | 5 | 764 | |||
NM_206894.3 | NP_996777.2 | 5 | 456 | |||
XM_005258903.4 | XP_005258960.1 | 5 | 823 | |||
XM_011526950.2 | XP_011525252.1 | 5 | 904 | |||
GenBank mRNA | AK297474.1 | - | 5 | 465 | ||
BC057245.1 | - | 5 | 458 | 6712908 | ||
BM473790.1 | - | 3 | 296 | |||
DA819130.1 | - | 5 | 454 | |||
DC308617.1 | - | 5 | 498 | |||
EL735861.1 | - | 3 | 335 |