Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00493 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015432.1 | 2 | 300 | |||
GenBank mRNA | AF147354.1 | - | 2 | 278 | ||
AK024594.1 | - | 2 | 300 | |||
BC015983.1 | - | 2 | 295 | |||
BC039422.1 | - | 2 | 279 | |||
BC092494.1 | - | 2 | 301 | |||
BC127753.1 | - | 2 | 279 | |||
BC127754.1 | - | 2 | 279 | |||
BC141963.1 | - | 2 | 279 | |||
BC141964.1 | - | 2 | 279 | |||
BC150559.1 | - | 2 | 282 | |||
BC150560.1 | - | 2 | 282 |