Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002281.3 | NP_002272.2 | 9 | 1460 | ||
GenBank mRNA | BC006452.1 | - | 5 | 628 | ||
BC021241.2 | - | 4 | 502 | |||
BC117465.1 | - | 10 | 1431 | 40126016 | ||
BC117467.1 | - | 10 | 1432 | 40126018 | ||
X80197.1 | - | 5 | 668 | |||
X81420.1 | - | 10 | 1172 |